Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(-)-Dibenzoyl-L-tartaric acid 98%

Supplier: Thermo Scientific Chemicals
Warning

43812-1000 43812-5000 43812-0250
TS43812-1000EA 94 USD InStock
TS43812-1000 TS43812-5000 TS43812-0250
(-)-Dibenzoyl-L-tartaric acid 98%
(-)-Dibenzoyl-L-tartaric acid


Formula: C₁₈H₁₄O₈
MW: 358.31
Melting Pt: 150…151.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00003074
CAS Number: 2743-38-6
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤1.0 %
HPLC ≥97.5 %
Specific optical rotation -114.0° to -117.0° (20°C, 589 nm) (c=9, ethanol)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty