Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-ethylenediamine 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: t-boc-N-EDAtert-Butyl N-(2-Aminoethyl)carbamateN-(tert-Butoxycarbonyl)-1,2-ethylenediaminetert-Butyl (2-aminoethyl)carbamateN-(tert-Butoxycarbonyl)-1,2-diaminoethaneN-BOC-1,2-diaminoethane

38990-0010 38990-0050 38990-0250
TS38990-0050EA 317 USD InStock
TS38990-0050 TS38990-0250 TS38990-0010
N-Boc-ethylenediamine 98%
N-Boc-ethylenediamine


Formula: C₇H₁₆N₂O₂
MW: 160.22 g/mol
Boiling Pt: 80…84 °C (.3 mmHg)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
UN: 2735
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4550 to 1.4570 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty