Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Amino-1-propanesulphonic acid 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Homotaurine3-Aminopropane-1-sulfonic acidTramiprosate3APS3-Aminopropane-1-sulphonic acid

10443-0050 10443-0010
TS10443-0010EA 38 USD InStock
TS10443-0010 TS10443-0050
3-Amino-1-propanesulphonic acid 95%
3-Amino-1-propanesulfonic acid


Formula: C₃H₉NO₃S
MW: 139.17 g/mol
Melting Pt: >270 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008225
CAS Number: 3687-18-1
Order Now
Specification Test Results
Appearance White to light blue Crystalline powder
Infrared spectrum Conforms
Melting point ≥270 °C
Titration with NaOH ≥94.0 %
Phosphate (PO4) ≤3.5 % (by acid extraction)
Calcium (Ca) ≤200 ppm (by acid extraction)
Iron (Fe) ≤150 ppm (by acid extraction)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty