Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-ɑ-bromophenylacetate 97+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: ɑ-Bromophenylacetic Acid Methyl EsterMethyl alpha-broMophenylacetate

37930-0050 37930-0250
TS37930-0250EA 280 USD InStock
TS37930-0250 TS37930-0050
Methyl-ɑ-bromophenylacetate 97+%
Methyl-alpha-bromophenylacetate


Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 113 °C (3 mmHg)
Density: 1.455 g/cm³
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013535
CAS Number: 3042-81-7
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥97.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty