Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfo-SANPAH (Sulfosuccinimidyl-6-(4′-azido-2′-nitrophenylamino)hexanoate), No-Weigh™ Format, Pierce™

Supplier: 0000013509
Warning
Synonyms: Sulfo SANPAH

A35395
103523-944EA 288 USD InStock
103523-944
Sulfo-SANPAH (Sulfosuccinimidyl-6-(4′-azido-2′-nitrophenylamino)hexanoate), No-Weigh™ Format, Pierce™
Sulfo-SANPAH (Sulfosuccinimidyl-6-(4′-azido-2′-nitrophenylamino)hexanoate)
Thermo Scientific Pierce Sulfo-SANPAH is a hetero-bifunctional crosslinker that contains an amine-reactive N-hydroxysuccinimide (NHS) ester and a photoactivatable nitrophenyl azide.

  • Reactive groups: sulfo-NHS ester and nitrophenyl azide
  • Reactive towards: amino groups and any nucleophile
  • Non-cleavable
  • N-Sulfosuccinimidyl-6-(4'-azido-2'-nitrophenylamino) hexanoate
  • Optimal photolysis occurs at 320-350 nm; minimizes damage to biomolecules by irradiation
  • Water soluble; non-cleavable

NHS esters react efficiently with primary amino groups (-NH2) in pH 7–9 buffers to form stable amide bonds. The reaction results in the release of sulfo-N-hydroxy-succinimide. When exposed to UV light, nitrophenyl azides form a nitrene group that can initiate addition reactions with double bonds, insertion into C-H and N-H sites, or subsequent ring expansion to react with a nucleophile (e.g., primary amines). Sulfo-SANPAH, supplied as a sodium salt, is useful for cell-surface protein crosslinking and possesses charged groups for water solubility to a concentration of 10 mM.

Thermo Scientific No-Weigh products are specialty reagents provided in a pre-aliquoted format. The pre-weighed packaging prevents the loss of reagent reactivity and contamination over time by eliminating the repetitive opening and closing of the vial. The format enables use of a fresh vial of reagent each time, eliminating the hassle of weighing small amounts of reagents and reducing concerns over reagent stability.

Caution: For Research Use Only. Not for use in diagnostic procedures.


Formula: C₁₆H₁₇O₉N₆NaS
MW: 492.40 g/mol
Storage Temperature: Freezer
CAS Number: 102568-43-4
Order Now
Specification Test Results
Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Sulfo-NHS Ester, Aryl Azide
Spacer Long
Form Solid
Quantity 50 mg
Type Crosslinker
Solubility Water

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty