Would you like to log in again?
Pentair Hardness/Iron Reduction Double-Open-End Liquid Filter Cartridges
Supplier: Pentair
155251-43
MFLX29830-60EA 58.5 USD InStock
MFLX29830-60
Pentair Hardness/Iron Reduction Double-Open-End Liquid Filter Cartridges
Filters Filter Cartridges
Prevents scale and rust buildup—ideal for water processing.
  • Constructed with Hexametaphosphate crystals
  • FDA-compliant
Hexametaphosphate Crystal Cartridge prevents scale and rust buildup — use with water heaters, air conditioning equipment, and many other types of water processing equipment. Food-grade hexametaphosphate dissolves slowly in water to treat scale, corrosion, and iron problems at various flow rates and feed concentrations. All materials are FDA-compliant.
Water being treated should have no more than 257 mg/L hardness, with a pH range of 6.5 to 9.0. Maximum temperature is 100 °F (38 °C). Cartridge is made from polypropylene (PP) shell with a polyester pad prefilter and a PP post filter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty