Would you like to log in again?
Adapter Fittings, Compression to Female Threaded, Straight
Supplier: VWR International
25-4-2-K-O 25-6-6-K-PG 25-8-6-K-PG 25-5-4-K-PG 25-4-4-K-PG 25-6-4-K-PG
MFLX06384-76EA 37.9 USD InStock
MFLX06384-76 MFLX06384-71 MFLX06384-72 MFLX06384-73 MFLX06384-74 MFLX06384-75
Adapter Fittings, Compression to Female Threaded, Straight
Fittings and Connectors
Maintain a permanent connection with these adapters.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These threaded fittings are suitable for permanent installation and can used with your rigid tubing for high-pressure applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty