Would you like to log in again?
Saint Gobain® Plastic Sanitary Clamp to Single Hose Barb Adapters
Supplier: VWR International
Saint Gobain®
ATP250PPSB EA ATP500PPSB EA ATP375PSSB EA ATP250PFSB EA ATP250PSSB EA ATP500PFSB EA ATP375PPSB EA ATP375PFSB EA ATP500PSSB EA
MFLX31512-08EA 109 USD InStock
MFLX31512-08 MFLX31512-07 MFLX31512-06 MFLX31512-01 MFLX31512-00 MFLX31512-05 MFLX31512-04 MFLX31512-03 MFLX31512-02
Saint Gobain® Plastic Sanitary Clamp to Single Hose Barb Adapters
Fittings and Connectors
Quick and secure sanitary tubing attachments are critical in numerous low-pressure laboratory, biotechnology, and pharmaceutical applications. Contamination of fluids and product loss through leakage can be costly. These Sanitary Fittings provide a superior connection to tubing through a single-barb construction, reducing the potential for leakage.
  • Tapered funnel design reduces 'dead space' for exceptionally smooth fluid flow
  • Smooth inner surface prevents particle entrapment
  • Seamless construction prevents contamination by eliminating leaks and areas of potential bacterial growth
  • Purchase alongside flexible tubing, ½" sanitary gasket, and a mini tri-clamp to complete the assembly
Fittings are available in Kynar (PVDF), polypropylene (PP), and polysulfone (PS). These materials are all radiation sterilizable. Sterilize by gamma irradiation. Produced from materials that meet USP Class VI.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty