Would you like to log in again?
Micropump Cavity-Style Gear Pump Heads for NEMA Type 56 C-face Motors
Supplier: Micropump
L20563 200037 L20563+83400
MFLX73007-00EA 5050 USD InStock
MFLX73007-00 MFLX73007-11 MFLX73007-02
Micropump Cavity-Style Gear Pump Heads for NEMA Type 56 C-face Motors
Pumps Pump Heads Gear Pump Heads
Smooth pulseless flow provides precise, leak-free delivery of fluids.
  • Compact size and easy to service
  • Cavity-style heads are designed to handle higher inlet pressures.
Magnetically driven, precision-geared cavity pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and a single seal keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of fluids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty