Would you like to log in again?
Two-Way PVC Ball Valves, Female NPT
Supplier: CALCO LTD
TJH-001 TJH-002 TJH-003
MFLX07387-62EA 47.4 USD InStock
MFLX07387-62 MFLX07387-42 MFLX07387-22
Two-Way PVC Ball Valves, Female NPT
Valves Ball Valves
Create a secure pipe threaded connection.
  • Constructed of durable PVC
Control flow with the 90° rotating handle. Valves have seals made of Viton® fluoroelastomer and seats made of PTFE. Maximum pressure is 150 psi at 32 °F (10.3 bar at 0 °C); maximum temperature is 140 °F (60 °C at 1.7 bar).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty