Would you like to log in again?
Brass Female/Male Mini Manual Ball Valves
Supplier: Parker
MV608-2 MV608-6 MV608-4
MFLX30533-20EA 25.6 USD InStock
MFLX30533-20 MFLX30533-22 MFLX30533-24
Brass Female/Male Mini Manual Ball Valves
Valves Ball Valves
Don’t worry about leaks.
  • Full flow design ensures maximum operating efficiency
  • Blow-out proof stems
Mini brass ball valves are designed for use in confined and hard-to-reach areas and are ideal for use on water and air service lines.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty