Would you like to log in again?
Cast iron float and thermostatic steam traps (parallel configuration)
Supplier: Spirax Sarco
FT-125 3/4" 50932 FT-15 3/4" 50926 FT-75 3/4" 50930 FT-30 3/4" 50928
MFLX07600-32EA 493 USD InStock
MFLX07600-32 MFLX07600-30 MFLX07600-36 MFLX07600-34
Cast iron float and thermostatic steam traps (parallel configuration)
Filters Gas Filters
Keep steam space free of condensate to provide maximum system efficiency.
  • Constructed of durable cast iron
Float and thermostatic steam traps contain a float valve mechanism which modulates to discharge condensate continuously at steam temperature, while non-condensable gases are released by a separate internal balanced pressure thermosatic air vent. Stainless steel internal parts can be serviced without disturbing the piping connections.
Typical applications:
All process equipment, particularly when controlled by modulating temperature control valves
Unit heaters
Air heat coils
Heat exchangers
Steam main drip stations
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty