Would you like to log in again?
Carbon Block Cartridges for "Big Blue" Housing
Supplier: Big Blue
155548-43 155170-43 155583-43 155312-43
MFLX01512-56EA 123 USD InStock
MFLX01512-56 MFLX01508-98 MFLX01512-54 MFLX01508-89
Carbon Block Cartridges for "Big Blue" Housing
Filters Filter Cartridges
Remove organic tastes and odors, chlorine and sediment.
  • Constructed with bonded powdered activated carbon
The bonded powdered activated carbon filter uses a design to remove organic tastes and odors, chlorine and sediment. Ideal for low or highly variable flow applications since flow cannot channel through the media like granular carbon cartridges††. Cryptosporidium and Giardia cysts are also removed with the 0.5-micron design.
Cartridges are made from FDA compliant materials and meet NSF Standard 42 for materials.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty