Would you like to log in again?
Low-Cost Battery-Operated Turbine Flowmeters/Totalizers
Supplier: Great Plains Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
TM15SQ9GMB TM050-P TM05NQ9GMA TM10NQ9GMA DISCONTINUED TM400 TM10SQ9GMA TM07SQ9GMA TM05SQ9GMA TM20NQ9GMB TM050-N-P TM20SQ9GMB TM075-P TM07NQ9GMA TM15NQ9GMB TM100-P
MFLX05611-51EA 437 USD InStock
MFLX05611-51 MFLX05611-50 MFLX05611-31 MFLX05611-52 MFLX05611-19 MFLX05611-18 MFLX05611-15 MFLX05611-14 MFLX05611-17 MFLX05611-16 MFLX05611-33 MFLX05611-11 MFLX05611-10 MFLX05611-54 MFLX05611-13 MFLX05611-12
Low-Cost Battery-Operated Turbine Flowmeters/Totalizers
Flow Meters
Local display includes rate of flow, batch, and cumulative totals.
  • Heavy-duty schedule 80 PVC construction
These durable meters offer accuracy at a reasonable price for measuring water and are available in flow ranges from 1 to 200 GPM and 1/2'' to 2'' inch fitting sizes. Meters indicate totalization and rate of flow, and read up to 999,999 gallons or liters — switch between gallon and liter and the electronics will indicate accurately in both units of measure. Models 05611-10 to -19 offer digital indication powered by two replaceable lithium batteries. Models 05611-50 to -59 have no display and have a square wave pulse output. Flowmeters are available with NPT or male spigot fittings, and work perfect for existing PVC plumbed systems. Wetted parts include the PVC housing, tungsten carbide shaft, PVDF rotor and supports and ceramic bearings. Flowmeters are field calibrateable.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty