Would you like to log in again?
Union Fittings, Hosebarb to Threaded Adaper, Straight, 316 Stainless Steel
Supplier: VWR International
NPL075-SS NPL050-SS NPL125-SS NPL300-SS NPL100-SS
MFLX33343-44EA 32.1 USD InStock
MFLX33343-44 MFLX33343-46 MFLX33343-40 MFLX33343-42 MFLX33343-54
Union Fittings, Hosebarb to Threaded Adaper, Straight, 316 Stainless Steel
Fittings and Connectors
Quick and easy assembly without tools.
  • Constructed of durable stainless steel
316 Stainless Steel Combination Connection Nipples have NPT threads. Recommended for low-pressure discharge and suction service. To tighten the hose onto the nipple use the 33350-series band clamps (sold separately).
Steel Hose Menders join two hoses together or repairs a piece of hose. Ideal for liquid applications — not recommended for compressible products such as air. Tighten hose to mender with band clamps (sold separately).
Double-Bolt Clamps are rugged general-purpose clamps made of plated iron and designed to tightly grip the hose. The inner surface has dual ridges and the bolt lugs are reinforced to prevent bending out of alighment. Simply match the size to the outside diameter of the hose.
Band Clamps are ideal for tightening hose over barbed fittings. Constructed of stainless steel. All band clamps require a band clamp tool to install.
Brass Push-On Fittings are for use with the Gates® Python hose only. These fittings are designed to grip and hold pressure when properly installed — fitting ridges dig into the hose wall to provide a tight, leak-free connection. Note: Do not use hose clamps with these fittings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty