Would you like to log in again?
Parker Hannifin Adapter Fittings, Compression Branch to Male Threaded, Tee
Supplier: Parker Hannifin
4MBT4N-316 GAMT-121212 GBMT-222 2MBT2N-316 GBMT-888 P4MT4 6MBT4N-316 GBMT-886 GBMT-224 GBMT-666 P10MT8 6MBT6N-316 GBMT-442 P6MT6 GBMT-664 GBMT-884 GBMT-668 GBMT-444
MFLX31321-81EA 97.6 USD InStock
MFLX31321-81 MFLX30486-65 MFLX30486-66 MFLX31321-80 MFLX30486-63 MFLX06444-76 MFLX30486-64 MFLX31320-87 MFLX31320-86 MFLX31320-38 MFLX31320-39 MFLX31320-36 MFLX31320-37 MFLX31320-34 MFLX31321-79 MFLX31320-35 MFLX06444-72 MFLX06444-71
Parker Hannifin Adapter Fittings, Compression Branch to Male Threaded, Tee
Fittings and Connectors
Securely connect your rigid tubing.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
These branch tees feature a male NPT thread on the bottom branch for quick connection to a threaded port.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty