Would you like to log in again?
Parker Hannifin Adapter Fittings, Compression to Female Threaded, Straight
Supplier: Parker Hannifin
GAFS-22 GAFS-812 GAFS-128 GAFS-1212 P4FC4 GAFS-66 P6FC8 GAFS-46 GAFS-84 GAFS-64 GAFS-68 GAFS-44 GAFS-86 GAFS-42 GAFS-48 GAFS-24 GAFS-88
MFLX06444-11EA 170 USD InStock
MFLX06444-11 MFLX06444-13 MFLX31320-54 MFLX31320-55 MFLX31320-74 MFLX31320-52 MFLX31320-53 MFLX31320-50 MFLX31320-72 MFLX31320-73 MFLX31320-51 MFLX31320-70 MFLX31321-82 MFLX31320-71 MFLX31320-58 MFLX31320-56 MFLX31320-57
Parker Hannifin Adapter Fittings, Compression to Female Threaded, Straight
Fittings and Connectors
Excellent choice to connect rigid tubing systems.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty