Would you like to log in again?
Female Cam to Male NPT Threaded Couplers, Polypropylene
Supplier: VWR International
QC-B075-PP QC-B200-PP QC-B150-PP
MFLX34020-36EA 23.9 USD InStock
MFLX34020-36 MFLX34020-35 MFLX34020-32
Female Cam to Male NPT Threaded Couplers, Polypropylene
Fittings and Connectors
Cam couplers provide quick disconnection from system.
  • All fittings have EPDM gaskets
  • Constructed of durable polypropylene
Cam couplers are ideal for providing a quick-disconnect solution to your pump line.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty