Would you like to log in again?
Great Plains Economy Fuel and Water Meters
Supplier: Great Plains Industries
113255-2 113255-5 113255-6 113255-3
MFLX32486-00EA 244 USD InStock
MFLX32486-00 MFLX32485-99 MFLX32486-02 MFLX32486-01
Great Plains Economy Fuel and Water Meters
Flow Meters
Small, powerful, and durable.
  • Battery powered 4 digit display
  • Factory calibrated to water or fuel
  • Gallon or litre measurement units
  • Batch and cumulative totals
  • Ready to install
Small, powerful, microprocessor based totalizers powered by AAA batteries have large four-digit displays and mount at the nozzle for ultimate convenience. They are suited for flow ranges from 3 to 30 GPM. Nylon® models, rated to 150 PSIG, are factory calibrated for water. Aluminum models, rated to 300 PSIG are factory calibrated for fuels. Accuracy is ±5%. All mount with 1 inch female NPT fittings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty