Would you like to log in again?
Extra-Strength Pleated Cellulose Cartridges
Supplier: PENTAIR RESIDENTIAL FILTRATION LLC
255481-43 255482-43
MFLX01509-11EA 15.5 USD InStock
MFLX01509-11 MFLX01509-09
Extra-Strength Pleated Cellulose Cartridges
Filters Filter Cartridges
Pleated filter gives you more surface area for higher flowrate and longer life.
  • Constructed of cellulose and polyester
This cellulose cartridge is blended with polyester for better wet strength. The polyester material allows more pleats to improve flow and dirt loading. Suitable to 165 °F (74 °C).
Note: For optimal performance, change cartridge at 25 psid.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty