Would you like to log in again?
Wika LS-10 Submersible Level Transmitters with Various Cable Lengths
Supplier: Wika
50304003 50043561 50133179 50168185 50450701 50162322 50597680 4262809-0009 50179802 50211633 50187813 52820507 50390473 4275466 50380982 50303953
MFLX30004-60EA 1508 USD InStock
MFLX30004-60 MFLX30004-26 MFLX30004-15 MFLX30004-03 MFLX30004-14 MFLX30004-02 MFLX30004-01 MFLX30004-33 MFLX30004-22 MFLX30004-32 MFLX30004-21 MFLX30004-20 MFLX30004-08 MFLX30004-39 MFLX30004-27 MFLX30004-16
Wika LS-10 Submersible Level Transmitters with Various Cable Lengths
Level Sensors
Compact, rugged submersible pressure transmitter.
  • Constructed of 316 stainless steel (SS), polyurethane, and polyamid
WIKA submersible liquid level transmitters and transducers are engineered for a wide variety of industrial and municipal liquid level measurement applications. Each transmitter undergoes extensive quality control testing and calibration to achieve an accuracy <0,25% full scale. The printed circuit boards use state-of-the-art surface mount technology for protection against mechanical shock and vibration. Each is temperature compensated to ensure accuracy and longterm stability when exposed to severe ambient temperature variations. The transmitter features a watertight, vented polyurethane cable that can withstand over 220 pounds of strain. This allows the transmitter to be supported without any additional cabling. The transmitter meets NEMA 6P and IP 68 requirements for submersion up to 300 feet. This compact, rugged submersible pressure transmitter is suitable for applications in level measurement, water and wastewater treatment, well depth measurement, and offshore water depth measurement. Order transmitter with cable length required for your application below. Custom cable lengths available.
Note: Intrinsically Safe versions available
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty