Would you like to log in again?
Pentair Quick-Change Filtration System Accessories
Supplier: Pentair
257438-06 155962-43
MFLX01507-42EA 48.5 USD InStock
MFLX01507-42 MFLX01507-90
Pentair Quick-Change Filtration System Accessories
Filters Cartridge Filter Systems
Optimize your Pentair Quick-Change Filtration Systems.
  • Choose from replacement cartridges and bracketed heads for your your Pentair Quick-Change Filtration Systems
The quick-change filtration systems offer a complete solution in one simple design—several cartridges will meet your water quality needs. Easily change the cartridge filter with a quarter-turn of the filter. The filter head features a built-in auto shut-off when cartridge is removed, eliminating the need for additional shutoff valves in your filter system. Connections on inlets and outlets are available with 1/4" NPT fittings, or 1/4" and 3/8" John Guest® push-to-connect fittings.
Each quick-change system includes a cartridge, filter head, and mounting bracket for use as a single unit; single cartridge systems are NSF certified. You can also combine cartridges in a series of up to three filters with the optional double and triple bracketed heads (sold separately).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty