Would you like to log in again?
Stenner Compact Economical Peristaltic Pumps
Supplier: Stenner
E20T4H71S8 E10LHMA1S6 E10LHFA1S6 E20T4G71S8 E20T4F71S8 E10LHFC5SWE
MFLX74209-00EA 640 USD InStock
MFLX74209-00 MFLX74209-03 MFLX74209-02 MFLX74209-05 MFLX74209-04 MFLX74209-06
Stenner Compact Economical Peristaltic Pumps
Pumps Peristaltic Pumps
Effortless tube replacement with quick-release pump head – no tools required.
  • Tubing is bonded to the fittings during manufacturing for a strong, single-piece tubing assembly
  • Run dry and self-prime without worrying about tubing or pump damage
  • Enclosed housing is wall mountable (mounting stand sold separately)
These Stenner compact economical peristaltic pumps can be run dry and are self-priming for easy pumping. Pumps are available in two models—timed and low-volume models.
Delivery information: Includes two pump tubes, three 1/4" connecting nuts and ferrules, injection valve/injection fitting, weighted suction line strainer, 20'' (6 m) of 1/4" PE suction/discharge tubing, and AC power adapter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty