Would you like to log in again?
Ward's® Live Waxworms (Galleria mellonella)
Supplier: VWR
Ward's®
6WW250 18WW36 572-0058-9 WARD470184-798
470338-856EA 10.7 USD InStock
470338-856 470184-798 470336-808 470336-812
Ward's® Live Waxworms (Galleria mellonella)
Specimens Live Specimens Animals - Living Material
Wax worms are a hardy larvae that are easy to handle.
  • Chemical-free, safe for feeding to pets
  • Healthy for many reptiles and fish
  • These creamy white worms have soft, grublike bodies and are dry to the touch
  • Refrigeration optional
Wax worms do not need moisture, and you do not need to feed them as in their larva state they live off their own body fat. You can keep them at room temperature, or around 45 to 65 °F for maximum shelf life.
Delivery information: These products contain living materials and will ship via overnight or 2 nd day delivery. Please specify a delivery date during checkout.
Packaging: Packed in cups with wood chips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty