Would you like to log in again?
LED Photo-Crosslinking Device, AlphaThera
Supplier: ALPHA THERA, LLC MS
AT8001-D
MSPP-AT8001-DEA 607.69 USD InStock
MSPP-AT8001-D
LED Photo-Crosslinking Device, AlphaThera
UV Crosslinkers
AlphaThera’s LED Photo-Crosslinking Device (LEDPX) uses non-damaging light (365 nm) to enable the efficient and site-specific labeling of antibodies with all oYo-Link products.
Two LED lamps are used to provide a uniform intensity UV exposure, while generating less heat than a fluorescent bulb.
The LEDPX includes a digital countdown timer, which can be set to one or two hours illumination. There is an LED display that continuously indicates the amount of time remaining. There is also a cooling fan that can be turned on/off, to reduce sample heating.
The device has a small footprint, a pull-out sample tray, and includes an acrylic container for placing samples in the device.
Note: oYo-Link products are compatible with a wide range of long-wavelength (365 nm) UV photo-crosslinking devices. Suitable alternatives include UVP UVL-56 and UVP CL-1000L.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty