Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromoform 96% stabilized

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Tribromomethane

15820-2500 15820-0050 15820-0025 15820-0010
TS15820-2500EA 206 USD InStock
TS15820-2500 TS15820-0025 TS15820-0010 TS15820-0050
Bromoform 96% stabilized
Bromoform


Formula: Br₃CH
MW: 252.73 g/mol
Boiling Pt: 150…151 °C (760 mmHg)
Melting Pt: 8 °C
Density: 2.63 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000128
CAS Number: 75-25-2
EINECS: 200-854-6
UN: 2515
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/78/85/717885.jpg
Order Now
Specification Test Results
Appearance Clear colorless to yellow to light orange Liquid
Infrared spectrum Conforms
GC ≥95 %
Stabilizer 3 to 5 wt% ethanol
Mercury (Hg) ≤0.05 ppm
Insoluble matter ≤0.2 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty