Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromoform 99+% stabilized

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Tribromomethane

22069-5000 22069-1000 22069-0250
TS22069-5000EA 728.57 USD InStock
TS22069-5000 TS22069-0250 TS22069-1000
Bromoform 99+% stabilized
Bromoform


Formula: Br₃CH
MW: 252.73 g/mol
Boiling Pt: 150…151 °C (760 mmHg)
Melting Pt: 8 °C
Density: 2.89 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000128
CAS Number: 75-25-2
EINECS: 200-854-6
UN: 2515
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/78/85/717885.jpg
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥99.0 % (excluding stabilizer)
Stabilizer ≤1.5 % w/w ethanol

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty