Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloro-2-pentanone 90%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Chloropentan-2-one

38829-1000 38829-0250
TS38829-0250EA 48 USD InStock
TS38829-0250 TS38829-1000
5-Chloro-2-pentanone 90%
5-Chloro-2-pentanone


Formula: C₅H₉ClO
MW: 120.58 g/mol
Boiling Pt: 172…175 °C (760 mmHg)
Melting Pt: –48 °C
Density: 1.053 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00001008
CAS Number: 5891-21-4
EINECS: 227-565-8
UN: 1224
ADR: 3,III
Order Now
Specification Test Results
Appearance Brown to black Liquid
Infrared spectrum Conforms
GC ≥88 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty