Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Chloroadamantane 98%

Supplier: Thermo Scientific Chemicals
Warning

26424-2500 26424-0500 26424-0100
TS26424-0100EA 87 USD InStock
TS26424-0100 TS26424-0500 TS26424-2500
1-Chloroadamantane 98%
1-Chloroadamantane


Formula: C₁₀H₁₅Cl
MW: 170.68 g/mol
Melting Pt: 163…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00075627
CAS Number: 935-56-8
EINECS: 213-305-0
Order Now
Specification Test Results
Appearance White to light grey Adhering crystalline solid and/or chunks
Infrared spectrum Conforms
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty