Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Pentyn-1-ol 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Pentyn-5-ol

33043-0250 33043-0050
TS33043-0050EA 55 USD InStock
TS33043-0050 TS33043-0250
4-Pentyn-1-ol 95%
4-Pentyn-1-ol


Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 154…155 °C
Density: 0.91 g/cm³
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00002974
CAS Number: 5390-04-5
EINECS: 226-383-6
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4430 to 1.4470 (20°C, 589 nm)
GC ≥94 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty