Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Dinitrobenzoyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

40871-0250 40871-1000 40871-5000
TS40871-1000EA 120 USD InStock
TS40871-1000 TS40871-0250 TS40871-5000
3,5-Dinitrobenzoyl chloride 99%
3,5-Dinitrobenzoyl chloride


Formula: C₇H₃ClN₂O₅
MW: 230.56 g/mol
Boiling Pt: 196 °C (15hPa)
Melting Pt: 67…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00007248
CAS Number: 99-33-2
EINECS: 202-750-6
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance Yellow to beige Crystals or crystalline needles
Infrared spectrum Conforms
Melting point 67 to 71 °C
Titration RCOCl ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty