Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrachloro-p-benzoquinone 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Tetrachloroquinonep-Chloranil2,3,5,6-Tetrachlorobenzo-1,4-quinone2,3,5,6-Tetrachloro-1,4-benzoquinoneTetrachloro-1,4-benzoquinone4-Chloranil

21356-2500 21356-5000 21356-1000
TS21356-5000EA 479 USD InStock
TS21356-5000 TS21356-2500 TS21356-1000
Tetrachloro-p-benzoquinone 99%
Tetrachloro-p-benzoquinone


Formula: C₆Cl₄O₂
MW: 245.88 g/mol
Melting Pt: >290 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00001594
CAS Number: 118-75-2
EINECS: 204-274-4
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Yellow to yellow-green Powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C, 3 hrs)
HPLC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty