Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) 98% free radical

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2,2,6,6-Tetramethyl-4-hydroxy-1-piperidinyloxy radicalTempol4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-Oxyl

17505-0250 17505-0050 17505-0010
TS17505-0250EA 311.07 USD InStock
TS17505-0250 TS17505-0010 TS17505-0050
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) 98% free radical
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-Hydroxy-TEMPO, TEMPOL, free radical)


Formula: C₉H₁₈NO₂
MW: 172.25 g/mol
Melting Pt: 69…73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006478
CAS Number: 2226-96-2
EINECS: 218-760-9
Order Now
Specification Test Results
Appearance Orange Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 69 to 73 °C
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty