Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diphenylphosphoryl azide 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Diphenylphosphonic azideDiphenyl azidophosphate

40904-1000 40904-5000 40904-0250
TS40904-1000EA 235 USD InStock
TS40904-1000 TS40904-0250 TS40904-5000
Diphenylphosphoryl azide 98%
Diphenylphosphoryl azide


Formula: C₁₂H₁₀N₃O₃P
Boiling Pt: 157 °C (.2 mmHg)
Density: 1.273 g/cm³
Flash Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001987
CAS Number: 26386-88-9
EINECS: 247-644-0
UN: 3278
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear yellow to amber Liquid
Infrared spectrum Conforms
Refractive index 1.5500 to 1.5520 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty