Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Piperazinone 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Piperazin-2-one

39871-0010 39871-0050
TS39871-0010EA 82 USD InStock
TS39871-0010 TS39871-0050
2-Piperazinone 95%
2-Piperazinone


Formula: C₄H₈N₂O
MW: 100.12 g/mol
Melting Pt: 132…135 °C
Storage Temperature: Ambient
MDL Number: MFCD01318687
CAS Number: 5625-67-2
Order Now
Specification Test Results
Appearance White to tan Crystalline powder or solid
Infrared spectrum Conforms
GC ≥94 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty