Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Bromo-2-methylindole 98%

Supplier: Thermo Scientific Chemicals
Warning

43756-0050 43756-0010
TS43756-0050EA 267 USD InStock
TS43756-0050 TS43756-0010
5-Bromo-2-methylindole 98%
5-Bromo-2-methylindole


Formula: C₉H₈BrN
Melting Pt: 104…106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01863677
CAS Number: 1075-34-9
Order Now
Specification Test Results
Appearance Yellow to pink to tan Powder
Infrared spectrum Conforms
Melting point 103 to 107 °C
HPLC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty