Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone-p-toluenesulphonate 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Toluene-4-sulfonic acid (S)-5-oxo-pyrrolidin-2-ylmethyl ester(R)-(5-Oxopyrrolidin-2-yl)methyl 4-methylbenzenesulfonate(-)-L-Pyroglutamol p-toluenesulfonate(-)-L-Pyroglutamol p-toluenesulphonate(S)-5-(Hydroxymethyl)pyrrolidin-2-one 4-methylbenzenesulfonate(S)-5-(Hydroxymethyl)pyrrolidin-2-one 4-methylbenzenesulphonateToluene-4-sulphonic acid (S)-5-oxo-pyrrolidin-2-ylmethyl ester(R)-(5-Oxopyrrolidin-2-yl)methyl 4-methylbenzenesulphonate

34841-0010 34841-0050 34841-0250
TS34841-0250EA 552 USD InStock
TS34841-0250 TS34841-0010 TS34841-0050
(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone-p-toluenesulphonate 95%
(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone-p-toluenesulfonate


Formula: C₁₂H₁₅NO₄S
MW: 269.32 g/mol
Melting Pt: 122…129 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00274229
CAS Number: 51693-17-5
Order Now
Specification Test Results
Appearance Colorless to beige Powder
Infrared spectrum Conforms
Melting point 122 to 129 °C
HPLC ≥94 %
Specific optical rotation +8° to +9° (25 °C, 589 nm) (c=1, C2H5OH)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty