Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

(R)-2-Amino-7-hydroxytetralin 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (7R)-7-Amino-5,6,7,8-tetrahydro-2-naphthalenol(7R)-7-Amino-5,6,7,8-tetrahydronaphthalen-2-ol(3R)-3-aminotetralin-6-ol

43558-0050 43558-0010
TS43558-0010EA 184 USD InStock
TS43558-0010 TS43558-0050
(R)-2-Amino-7-hydroxytetralin 97%
(R)-2-Amino-7-hydroxytetralin


Formula: C₁₀H₁₃NO
MW: 163.22 g/mol
Melting Pt: 143.4…146.3 °C
Storage Temperature: Ambient
MDL Number: MFCD11617807
CAS Number: 85951-61-7
Order Now
Specification Test Results
Appearance White to grey Crystalline powder
Infrared spectrum Conforms
Melting point 143 to 151 °C
Specific optical rotation +85° to +95° (20 °C, 589 nm) (c=0.5, methanol)
Titration with HClO4 96.0 to 104.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty