Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromoheptane 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Heptyl bromide

10703-2500 10703-0010
TS10703-0010EA 218 USD InStock
TS10703-0010 TS10703-2500
1-Bromoheptane 99%
1-Bromoheptane


Formula: BrH₂C(CH₂)₅CH₃
MW: 179.1 g/mol
Boiling Pt: 180 °C (760 mmHg)
Melting Pt: –58 °C
Density: 1.14 g/cm³
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00000273
CAS Number: 629-04-9
EINECS: 211-068-8
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4489 to 1.4509 (20°C, 589 nm)
GC ≥98.5 %
Pore volume 0.75 to 0.80 ml/g
pH 6.5 to 7.0 (10 % suspension in water)
Volatile matter ≤6.0 % (160°C)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty