Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

9-Bromophenanthrene 96%

Supplier: Thermo Scientific Chemicals

10724-0250 10724-1000 10724-0100
TS10724-0250EA 71 USD InStock
TS10724-0250 TS10724-1000 TS10724-0100
9-Bromophenanthrene 96%
9-Bromophenanthrene


Formula: C₁₄H₉Br
MW: 257.13 g/mol
Boiling Pt: 180…190 °C (2 mmHg)
Melting Pt: 60…65 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001174
CAS Number: 573-17-1
EINECS: 209-351-6
Order Now
Specification Test Results
Appearance Light yellow Powder
Infrared spectrum Conforms
Melting point 60 to 65 °C
GC ≥95 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty