Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroacetonitrile 98+%

Supplier: Thermo Scientific Chemicals
Danger

10852-1000 10852-5000
TS10852-5000EA 163 USD InStock
TS10852-5000 TS10852-1000
Chloroacetonitrile 98+%
Chloroacetonitrile


Formula: C₂H₂ClN
MW: 75.5 g/mol
Boiling Pt: 124…126 °C (760 mmHg)
Melting Pt: –66 °C
Density: 1.189 g/cm³
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00001885
CAS Number: 107-14-2
EINECS: 203-467-0
UN: 2668
ADR: 6.1,I
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4240 (20°C, 589 nm)
GC ≥98.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty