Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromocyclopentane 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Cyclopentyl bromide

11160-2500 11160-0010 11160-0500
TS11160-2500EA 152.4 USD InStock
TS11160-2500 TS11160-0010 TS11160-0500
Bromocyclopentane 98%
Bromocyclopentane


Formula: C₅H₉Br
MW: 149.03 g/mol
Boiling Pt: 137…139 °C (760 mmHg)
Density: 1.39 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00001359
CAS Number: 137-43-9
EINECS: 205-294-6
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4881 to 1.4901 (20°C, 589 nm)
GC ≥97.5 %
UV (5 % in water)
UV at 210 nm A: ≤0.1
UV at 220 nm A: ≤0.07
UV at 230 nm A: ≤0.06
UV at 250 nm A: ≤0.03
UV at 260 nm A: ≤0.03
Titration after Ion exchange 98.5 to 101.5 % (on dry substance)
Chloride (Cl) ≤1000 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty