Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethyl phosphonate 98%

Supplier: Thermo Scientific Chemicals
Danger

11450-2500
TS11450-2500EA 53.91 USD InStock
TS11450-2500
Diethyl phosphonate 98%
Diethyl phosphonate


Formula: C₄H₁₁O₃P
MW: 138.1 g/mol
Boiling Pt: 60 °C (6mbar)
Melting Pt: –70 °C
Density: 1.08 g/cm³
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00044573
CAS Number: 762-04-9
EINECS: 212-091-6
Order Now
Specification Test Results
Appearance Colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4070 to 1.4090 (20°C, 589 nm)
GC ≥97.5 %
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty