Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitrobenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: p-Nitrobenzoyl chloride

15043-0050 15043-1000 15043-5000 15043-2500
TS15043-0050EA 40 USD InStock
TS15043-0050 TS15043-2500 TS15043-5000 TS15043-1000
4-Nitrobenzoyl chloride 98%
4-Nitrobenzoyl chloride


Formula: C₇H₄ClNO₃
MW: 185.57 g/mol
Boiling Pt: 202…205 °C (105 mmHg)
Melting Pt: 70…74 °C
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00007345
CAS Number: 122-04-3
EINECS: 204-517-4
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance Yellow Crystals or flakes
Infrared spectrum Conforms
Melting point 70 to 74 °C
Titration RCOCl ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty