Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Myricetin 95%

Supplier: Thermo Scientific Chemicals
Synonyms: 3,3',4',5,5',7-Hexahydroxyflavone3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one

15159-0250
TS15159-0250EA 104 USD InStock
TS15159-0250
Myricetin 95%
Myricetin


Formula: C₁₅H₁₀O₈
MW: 318.24 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006827
CAS Number: 529-44-2
Order Now
Specification Test Results
Appearance Brown-yellow to green-brown Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥94 %
HPLC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty