Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-5-bromovalerate 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Ethyl 5-bromovalerate5-Bromovaleric acid ethyl ester

15654-0250 15654-1000
TS15654-0250EA 79 USD InStock
TS15654-0250 TS15654-1000
Ethyl-5-bromovalerate 99%
Ethyl 5-bromovalerate


Formula: BrH₂C(CH₂)₃COOCH₂CH₃
MW: 209.08 g/mol
Boiling Pt: 104…109 °C (12 mmHg)
Density: 1.32 g/cm³
Flash Pt: 103 °C
MDL Number: MFCD00000266
CAS Number: 14660-52-7
EINECS: 238-705-2
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4570 to 1.4600 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty