Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chlorobenzothiazole 99%

Supplier: Thermo Scientific Chemicals
Warning

17349-0050 17349-0250
TS17349-0250EA 154 USD InStock
TS17349-0250 TS17349-0050
2-Chlorobenzothiazole 99%
2-Chlorobenzothiazole


Formula: C₇H₄ClNS
MW: 169.63 g/mol
Boiling Pt: 141 °C (1013 hPa)
Melting Pt: 21…24 °C
Density: 1.37 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD00005776
CAS Number: 615-20-3
EINECS: 210-415-0
Order Now
Specification Test Results
Appearance Clear yellow to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.6367 to 1.6387 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty