Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methylcyclohexanone 98%

Supplier: Thermo Scientific Chemicals
Warning

17426-0250 17426-1000
TS17426-1000EA 93 USD InStock
TS17426-1000 TS17426-0250
4-Methylcyclohexanone 98%
4-Methylcyclohexanone


Formula: C₇H₁₂O
MW: 112.17 g/mol
Boiling Pt: 169…171 °C (760 mmHg)
Melting Pt: –41 °C
Density: 0.914 g/cm³
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00001643
CAS Number: 589-92-4
EINECS: 209-665-3
UN: 2297
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4445 to 1.4465 (20°C, 589 nm)
GC ≥97.5 %
HPLC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty