Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

cis-1,4-Dichloro-2-butene 95%

Supplier: Thermo Scientific Chemicals
Danger

18760-1000 18760-0250
TS18760-1000EA 468 USD InStock
TS18760-1000 TS18760-0250
cis-1,4-Dichloro-2-butene 95%
cis-1,4-Dichloro-2-butene


Formula: C₄H₆Cl₂
MW: 125 g/mol
Boiling Pt: 152 °C (758 mmHg)
Melting Pt: –48 °C
Density: 1.18 g/cm³
Flash Pt: 55 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00062950
CAS Number: 1476-11-5
EINECS: 216-021-5
UN: 3489
ADR: 6.1,
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4884 to 1.4914 (20°C, 589 nm)
GC ≥94 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty