Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diphenylphosphinic chloride 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Diphenylphosphinyl chloride

21296-0500 21296-0100
TS21296-0500EA 232 USD InStock
TS21296-0500 TS21296-0100
Diphenylphosphinic chloride 98%
Diphenylphosphinic chloride


Formula: C₁₂H₁₀ClOP
Boiling Pt: 222 °C (16 mmHg)
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00002077
CAS Number: 1499-21-4
EINECS: 216-107-2
UN: 2920
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6080 to 1.6120 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty